Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Dopamine D1R/DRD1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP267934 Shop All Novus Biologicals Products
Click to view available options
:
Quantity: 1 Kit; Host Species: Rat

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Rat Dopamine D1R/DRD1 ELISA Kit (Chemiluminescence) measures rat Dopamine D1R/DRD1 in serum, plasma and other biological fluids.

Specifications

Assay Range 3.13-200 ng/mL
Assay Sensitivity 1.88 ng/mL
Cross Reactivity This kit recognizes Rat DRD1 in samples. No significant cross-reactivity or interference between Rat DRD1 and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 1812
Gene Symbol DRD1
Host Species Rat
Quantity 1 Kit
Research Discipline GPCR
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Dopamine D1 R/DRD1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.