Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ADAL Human Recombinant Protein
SDP

Catalog No. p-7104548 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.1 mg
0.5mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-267 of Human ADAL The Recombinant Human ADAL Protein is derived from E. coli. The Recombinant Human ADAL Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-Page
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol 0.4M UREA
Gene ID (Entrez) 161823
Molecular Weight (g/mol) TMW: 32.7kDa
Purity >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Quantity 0.5mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias adenosine deaminase-like, adenosine deaminase-like protein, DKFZp313B2137, EC 3.5.4, EC 3.5.4.-, FLJ44620
Common Name ADAL
Conjugate Unlabeled
Recombinant Yes
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.