Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Adenosine Deaminase 2/CECR1 Human Recombinant Protein
SDP

Catalog No. p-7105247 Shop All Novus Biologicals Products
Click to view available options
Quantity:
0.05mg
0.25mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 30-511 of Human Adenosine Deaminase 2/CECR1 The Recombinant Human Adenosine Deaminase 2/CECR1 Protein is derived from Baculovirus. The Recombinant Human Adenosine Deaminase 2/CECR1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-Page
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 51816
Molecular Weight (g/mol) TMW: 56.9kDa
Purity >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Quantity 0.05mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias ADA2, adenosine deaminase CECR1, ADGFadenosine deaminase 2, cat eye syndrome chromosome region, candidate 1, Cat eye syndrome critical region protein 1, CECR2, EC 3.5.4.4, IDGFL
Common Name Adenosine Deaminase 2/CECR1
Conjugate Unlabeled
Recombinant Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.