Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TGF-beta 1 Mouse Recombinant Protein
SDP

Catalog No. NBP25192525 Shop All Novus Biologicals Products
Click to view available options
Quantity:

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 279-390 of Mouse TGF-beta 1 The Recombinant Mouse TGF-beta 1 Protein is derived from E. coli. The Recombinant Mouse TGF-beta 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration

0.25mg/mL

For Use With (Application)

SDS-Page

Formulation

20mM Tris-HCl (pH 8.0) containing 10% glycerol

Gene ID (Entrez)

7040

Molecular Weight (g/mol)

TMW: 15kDa

Purity

>85%, by SDS-PAGE

Quantity

0.25mg

Storage Requirements

Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.

Gene Alias

CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1

Common Name

TGF-beta 1

Conjugate

Unlabeled

Recombinant

Yes

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.