Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Replacement Light Bulbs

For use with Microchem SafTest Analyzers (Models I and II)

Supplier:  MP Biomedicals™ 07WS1024

 View more versions of this product

Catalog No. MP07WS1024


missing translation for 'QuantityLeft'
Add to Cart

Description

Description

For use with Microchem SafTest Analyzers (Models I and II)
Specifications

Specifications

Replacement Bulb
MicroChem I (Model with Integrated Printer)
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.