Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Replacement Light Bulbs

For use with Microchem SafTest Analyzers (Models I and II)

$99.00 - $215.00

Specifications

Product Type Replacement Bulb
Packaging Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
MP07WS1024
View Documents
MP Biomedicals™
07WS1024
MicroChem I (Model with Integrated Printer)
Each for $99.00
Add to Cart
 
MP07WS1023
View Documents
MP Biomedicals™
07WS1023
MicroChem II (Model with External Printer)
Each for $215.00
Add to Cart
 
Description

Description

For use with Microchem SafTest Analyzers (Models I and II)
Specifications

Specifications

Replacement Bulb
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.