Learn More
Restek™ FastPack Inlet Kits for Agilent GCs
Includes all parts needed for routine maintenance of injection ports and the entire system.
Supplier: Restek™ 21102
Description
- Economical - costs less than the sum of the individual parts
- Factory sealed Mylar bag prevents contamination of products from weeks in the lab
- Each bag includes five maintenance kits
- Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Splitless Gooseneck Inlet Maintenance Kit | |
washer, septa and liner | |
Agilent GC |