Learn More
Restek™ FastPack Inlet Kits for Agilent GCs
SDP
Includes all parts needed for routine maintenance of injection ports and the entire system.
$319.00 - $334.00
Specifications
For Use With Agilent GC
Includes washer, septa and liner
View More Specs
Products 3
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
06-718-416
SDP
View Documents
Restek™
21104
Split w/Wool Inlet Maintenance Kit washer, septa and liner
Pack of 5 for $329.00
missing translation for 'QuantityLeft'
Add to Cart
06-718-410
SDP
View Documents
Restek™
21101
Splitless Inlet Maintenance Kit washer, septa and liner
Pack of 5 for $319.00
missing translation for 'QuantityLeft'
Add to Cart
06-718-413
SDP
View Documents
Restek™
21102
Splitless Gooseneck Inlet Maintenance Kit washer, septa and liner
Pack of 5 for $334.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Economical - costs less than the sum of the individual parts
  • Factory sealed Mylar bag prevents contamination of products from weeks in the lab
  • Each bag includes five maintenance kits
  • Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Specifications
Agilent GC
washer, septa and liner
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.