Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Safety Goggles Box, Acrylic
SDPSCI_ED

Safely store your glasses or goggles.

Supplier:  United Scientific™ P70106

 View more versions of this product

Catalog No. S15762


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • Store goggles, glasses, or other small lab products using this clear acrylic box.
  • Can be used on a desktop or can be mounted on a wall
  • Includes a lid to keep the box free of dust or contamination
  • Available in a small (18 cm high) and large (25 cm high) size.
Specifications

Specifications

Uncoated
18 cm
16 cm
Clear
Acrylic
None
7 in.
6.2 in.
Clear
Eyewear Storage Box
Reusable
Acrylic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.