Learn More
Tocris Bioscience™ SCH 51344
Potent MTH1 inhibitor
$255.00 - $961.00
Specifications
Purity | >99% |
---|---|
Product Type | SCH 51344 |
Molecular Weight (g/mol) | 316.35 |
Inhibitors | SCH 51344 |
Description
Potent MTH1 inhibitor (Kd = 49 nM). Inhibits Ras-induced malignant transformation and increases α-actin promoter-driven CAT activity in Ras-transformed cells. Has no effect on Ras-induced ERK and JNK activation. Inhibits Ras-induced membrane ruffling in REF-52 fibroblasts and blocks anchorage-independent growth of Ras-transformed tumor cell lines. Also induces DNA damage in SW480 colon cancer cells.Specifications
>99% | |
SCH 51344 | |
316.35 | |
SCH 51344 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.