Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium-Calcium Exchanger Antibody (C2C12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB12028690.025ML
Description
Sodium-Calcium Exchanger Monoclonal specifically detects Sodium-Calcium Exchanger in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Insect, Kangaroo, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Sodium-Calcium Exchanger | |
Monoclonal | |
1 mg/ml | |
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
SLC8A3 | |
Purified canine cardiac sodium/calcium exchanger. | |
0.025 mL | |
Primary | |
Sodium Calcium Exchanger (C2C12) | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgM |
Western Blot | |
C2C12 | |
Unconjugated | |
Na(+)/Ca(2+)-exchange protein 3, Na+/Ca2+ exchanger 3, NCX3sodium/calcium exchanger SLC8A3, sodium/calcium exchanger 3, solute carrier family 8 (sodium/calcium exchanger), member 3, solute carrier family 8 (sodium-calcium exchanger), member 3 | |
Mouse | |
Affinity purified | |
RUO | |
6547 | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction