Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium-Calcium Exchanger Antibody (C2C12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$196.10
Specifications
Antigen | Sodium-Calcium Exchanger |
---|---|
Clone | C2C12 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
Applications | Western Blot |
Description
Sodium-Calcium Exchanger Monoclonal specifically detects Sodium-Calcium Exchanger in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Insect, Kangaroo, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Sodium-Calcium Exchanger | |
1 mg/ml | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
6547 | |
Purified canine cardiac sodium/calcium exchanger. | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
C2C12 | |
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
Monoclonal | |
Ascites | |
RUO | |
Na(+)/Ca(2+)-exchange protein 3, Na+/Ca2+ exchanger 3, NCX3sodium/calcium exchanger SLC8A3, sodium/calcium exchanger 3, solute carrier family 8 (sodium/calcium exchanger), member 3, solute carrier family 8 (sodium-calcium exchanger), member 3 | |
SLC8A3 | |
IgM | |
Affinity purified | |
Sodium Calcium Exchanger (C2C12) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title