Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon™ Two Way Tubing Stopcocks

For industrial use. Dynalon™ Two Way Tubing Stopcocks allow the transfer of liquids or gases through tubing by releasing the flow or blocking the flow.
Supplier: Dynalon™ 8860540060
Description
- Easy turn white PE valve for on/off with friction fit red PP barbed tubing attachments
- Operating temperature: PP/PE +5 up to +40 °C
- Resistant to compression max. 1 bar or 14.5 psi
- With mounting plate for easy installation
Specifications
Polypropylene, Polyethylene | |
For 5 to 7 mm tubing ID | |
4 mm | |
Red-White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction