Learn More
PIG™ Universal Poly Drum Funnel with Hinged Lid
SDP
Large-diameter funnel that covers entire drum top for clean, easy draining and pouring.
Supplier:  PIG™ DRM672RD
 View more versions of this product
Catalog No. 18-366-521
Add to Cart
Description
Description
  • Covers entire top of 55 gal. drum to minimize drips and spills
  • Allows access to 3/4 in. bung for use of liquid-level gauge
  • Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals
  • Level inside surface allows easy draining of containers, parts, or filters
  • Zigzag ridges prevent splashes and catch parts or tools
  • 4 in. sidewalls add splash protection
  • Hinged lid blocks out dirt and contaminants while still allowing quick access
  • Multiple lid color options allow you to segregate drums for safe, compliant filling
  • Fits any 55-gallon, tight-head steel or poly drum
Specifications
Specifications
Drum Funnel
29 in.
11 in.
Red
74 cm
28 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.