Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Uvula Tissue Slides
SDP

Catalog No. NB151710 Shop All Novus Biologicals Products
Click to view available options
:
5 Slides

Slides can be used with any antibody shown to be effective in formalin-fixed tissue using a standard immunostaining protocol.

In order to avoid false negative results from DNA or RNA sequences not being present in a 4 um cellular cross-section (NBP3-11873), 10 um tissue sections (NBP3-11874) are recommended. This product has not been evaluated for effectiveness in ISH or FISH applications.

Specifications

Quantity 5 Slides
Species Human
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Paraffin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.