Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
$1066.00 - $1305.86
Specifications
Color | Amber |
---|---|
Product Line | Wheaton™ CompletePAK |
Sterility | Sterile |
Includes | Vial, Stopper, and Seal |
Catalog Number | Mfr. No. | Volume (Metric) | Closure Type | Color (Seal) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Closure Type | Color (Seal) | Price | Quantity & Availability | |||
15-112-153
|
DWK Life Sciences
W990003CPA |
2 mL | Ultra Pure Sterile Serum Stopper | Red |
Case of 210 for $1,066.00
|
|
|||
15-112-151
|
DWK Life Sciences
W990001CPA |
2 mL | Omni Flex 3G Sterile Serum Stopper | Blue |
Case of 210 for $1,167.28
|
|
|||
15-112-154
|
DWK Life Sciences
W990004CPA |
2 mL | Omni Flex 3G Sterile Serum Stopper | Red |
Case of 210 for $1,167.28
|
|
|||
15-112-155
|
DWK Life Sciences
W990005CPA |
2 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Red |
Case of 210 for $1,199.26
|
|
|||
15-112-157
|
DWK Life Sciences
W990007CPA |
5 mL | OmniFlex 3G Sterile Serum Stopper | Blue |
Case of 200 for $1,220.58
|
|
|||
15-112-158
|
DWK Life Sciences
W990008CPA |
5 mL | Ultra Pure Sterile Serum Stopper | Red |
Case of 200 for $1,252.56
|
|
|||
15-112-161
|
DWK Life Sciences
W990011CPA |
5 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Red |
Case of 200 for $1,252.56
|
|
|||
15-112-162
|
DWK Life Sciences
W990012CPA |
10 mL | Ultra Pure Sterile Serum Stopper | Blue |
Case of 158 for $1,172.60
|
|
|||
15-112-165
|
DWK Life Sciences
W990015CPA |
10 mL | Ultra Pure Sterile Serum Stopper | Red |
Case of 158 for $1,172.60
|
|
|||
15-112-163
|
DWK Life Sciences
W990013CPA |
10 mL | OmniFlex 3G Sterile Serum Stopper | Blue |
Case of 158 for $1,273.88
|
|
|||
15-112-166
|
DWK Life Sciences
W990016CPA |
10 mL | OmniFlex 3G Sterile Serum Stopper | Red |
Case of 158 for $1,273.88
|
|
|||
15-112-164
|
DWK Life Sciences
W990014CPA |
10 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Blue |
Case of 158 for $1,305.86
|
|
|||
Description
- Products supplied ready-to-use
- Reduces lead times
- Focuses your efforts on core competencies
- Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
- Frees up valuable clean room space
- Limits number of suppliers required to provide complete packaging product
- Amber for photosensitive contents
- Validated processes
- Saves valuable time and money while maintaining a high quality product and streamlining process
- One source for total packaging solutions
Specifications
Amber | |
Wheaton™ CompletePAK | |
Sterile | |
Vial, Stopper, and Seal |