Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Activated Nucleotidase 1/CANT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$165.00 - $487.50
Specifications
Antigen | Calcium Activated Nucleotidase 1/CANT1 |
---|---|
Applications | Western Blot |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158875
![]() |
Novus Biologicals
NBP158875 |
100 ul |
Each for $487.50
|
|
|||||
NBP15887520
![]() |
Novus Biologicals
NBP15887520UL |
20 μL |
Each for $165.00
|
|
|||||
Description
Calcium Activated Nucleotidase 1/CANT1 Polyclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human samples. It is validated for Western Blot.Specifications
Calcium Activated Nucleotidase 1/CANT1 | |
Unconjugated | |
RUO | |
Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1 | |
CANT1 | |
IgG |
Western Blot | |
Rabbit | |
Q8WVQ1 | |
124583 | |
Synthetic peptides corresponding to CANT1(calcium activated nucleotidase 1) The peptide sequence was selected from the middle region of CANT1. Peptide sequence SPDFGDIAVSHVGAVVPTHGFSSFKFIPNTDDQIIVALKSEEDSGRVASY. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title