Learn More
Tocris Bioscience™ NPS 2143 hydrochloride
Selective calcium-sensing receptor (CaSR) antagonist; orally active calcilytic agent
$324.50 - $1356.00
Specifications
Purity | >99% |
---|---|
Product Type | NPS 2143 hydrochloride |
Molecular Weight (g/mol) | 445.38 |
Inhibitors | NPS 2143 hydrochloride |
Description
Selective Ca2+-sensing receptor antagonist. Blocks increases in cytoplasmic Ca2+ concentrations elicited by human Ca2+ receptors expressed in HEK293 cells (IC50 = 43 nM). Also stimulates parathyroid hormone secretion from bovine parathyroid cells (EC50 = 41 nM). Orally active calcilytic compound.Specifications
>99% | |
NPS 2143 hydrochloride | |
445.38 | |
NPS 2143 hydrochloride |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.